Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009399.3 | NP_033425.3 | 3 - 4 | 310 | 93 | |
GenBank mRNA | AF019046.1 | - | 3 - 4 | 305 | 93 | |
AK138990.1 | - | 3 - 4 | 316 | 93 | ||
BC019185.1 | - | 3 - 4 | 307 | 5025470 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |