Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009469.3 | NP_033495.2 | 14 - 15 | 1511 | 95 | |
GenBank mRNA | AF053756.1 | - | 14 - 15 | 1203 | 95 | |
AF072370.1 | - | 14 - 15 | 1194 | 95 | ||
AK122357.1 | - | 13 - 14 | 1013 | 95 | ||
AK148317.1 | - | 14 - 15 | 1486 | 95 | ||
AK154939.1 | - | 14 - 15 | 1528 | 95 | ||
AK156534.1 | - | 14 - 15 | 1488 | 95 | ||
BC053062.1 | - | 14 - 15 | 1171 | 95 | ||
BC057121.1 | - | 14 - 15 | 1511 | 6834534 | 95 | |
BC059835.1 | - | 14 - 15 | 1462 | 6406755 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |