Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163633.1 | NP_001157105.1 | 2 - 3 | 709 | 80 | |
NM_001163634.1 | NP_001157106.1 | 2 - 3 | 793 | 80 | ||
NM_009528.3 | NP_033554.3 | 2 - 3 | 672 | 80 | ||
GenBank mRNA | AK087499.1 | - | 2 - 3 | 710 | 80 | |
BC052018.1 | - | 2 - 3 | 433 | 6401357 | 80 | |
BC066003.1 | - | 2 - 3 | 793 | 5701652 | 80 | |
M89802.1 | - | 2 - 3 | 301 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |