Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163643.1 | NP_001157115.1 | 6 - 7 | 1254 | 75 | |
NM_009582.4 | NP_033608.3 | 4 - 5 | 1078 | 75 | ||
GenBank mRNA | AK029882.1 | - | 4 - 5 | 887 | 75 | |
AK034374.1 | - | 6 - 7 | 1070 | 75 | ||
AK166435.1 | - | 6 - 7 | 1174 | 75 | ||
BC047158.1 | - | 3 - 4 | 872 | 6494216 | 75 | |
BC057572.1 | - | 6 - 7 | 1078 | 6401321 | 75 | |
U14636.1 | - | 4 - 5 | 910 | 75 | ||
U23789.1 | - | 3 - 4 | 868 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |