Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serping1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009776.3 | NP_033906.2 | 6 - 7 | 1134 | 80 | |
GenBank mRNA | AF010254.1 | - | 6 - 7 | 1109 | 80 | |
AK144292.1 | - | 5 - 6 | 1094 | 80 | ||
AK167585.1 | - | 5 - 6 | 1071 | 80 | ||
BC002026.1 | - | 5 - 6 | 1065 | 3485810 | 80 | |
Y10386.1 | - | 6 - 7 | 1091 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |