Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286809.1 | NP_001273738.1 | 2 - 3 | 464 | 68 | |
NM_009792.3 | NP_033922.1 | 3 - 4 | 497 | 68 | ||
NM_177407.4 | NP_803126.1 | 14 - 15 | 1185 | 68 | ||
GenBank mRNA | AK083245.1 | - | 14 - 15 | 1186 | 68 | |
AK122410.2 | - | 14 - 15 | 1181 | 68 | ||
BB659027.1 | - | 2 - 3 | 420 | 68 | ||
BC031745.1 | - | 14 - 15 | 1166 | 4506005 | 68 | |
X14836.1 | - | 14 - 15 | 1169 | 68 | ||
X87142.1 | - | 3 - 4 | 497 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |