Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009875.4 | NP_034005.2 | 1 - 2 | 989 | 72 | |
GenBank mRNA | AK046676.1 | - | 1 - 2 | 724 | 72 | |
AK047669.1 | - | 1 - 2 | 990 | 72 | ||
AK050240.1 | - | 1 - 2 | 976 | 72 | ||
AK158411.1 | - | 1 - 2 | 969 | 72 | ||
AK162146.1 | - | 1 - 2 | 709 | 72 | ||
BC014296.1 | - | 1 - 2 | 972 | 4193493 | 72 | |
CT010382.1 | - | 1 - 2 | 470 | 72 | ||
U09968.1 | - | 1 - 2 | 470 | 72 | ||
U10440.1 | - | 1 - 2 | 687 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |