Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278041.1 | NP_001264970.1 | 11 - 12 | 1392 | 73 | |
NM_009893.2 | NP_034023.1 | 11 - 12 | 1326 | 73 | ||
GenBank mRNA | AF069501.1 | - | 11 - 12 | 1326 | 73 | |
AF096276.1 | - | 11 - 12 | 1305 | 73 | ||
AK077460.1 | - | 12 - 13 | 1428 | 73 | ||
AK149445.1 | - | 11 - 12 | 1301 | 73 | ||
AK165339.1 | - | 11 - 12 | 1319 | 73 | ||
BC145017.1 | - | 10 - 11 | 1533 | 73 | ||
BC145018.1 | - | 12 - 13 | 1454 | 73 | ||
BC145019.1 | - | 12 - 13 | 1432 | 73 | ||
BC145696.1 | - | 11 - 12 | 1390 | 40130999 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |