Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK037898.1 | - | 1 - 2 | 226 | 91 | |
AK134109.1 | - | 1 - 2 | 152 | 91 | ||
X86405.1 | - | 1 - 2 | 69 | 91 | ||
X93168.1 | - | 1 - 2 | 137 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mus musculus 1700 Inventoried Probe spans exons |