Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009971.1 | NP_034101.1 | 1 - 2 | 107 | 90 | |
GenBank mRNA | AK145177.1 | - | 1 - 2 | 77 | 90 | |
BC120759.1 | - | 1 - 2 | 60 | 40129682 | 90 | |
BC120761.1 | - | 1 - 2 | 60 | 40129684 | 90 | |
M13926.1 | - | 1 - 2 | 107 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |