Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162425.1 | NP_001155897.1 | 3 - 4 | 523 | 70 | |
NM_010107.4 | NP_034237.3 | 3 - 4 | 559 | 70 | ||
GenBank mRNA | AK009144.1 | - | 3 - 4 | 556 | 70 | |
AK079266.1 | - | 3 - 4 | 526 | 70 | ||
BC002046.1 | - | 3 - 4 | 524 | 3484518 | 70 | |
CN696019.1 | - | 3 - 4 | 559 | 70 | ||
U26188.1 | - | 3 - 4 | 518 | 70 | ||
U90662.1 | - | 3 - 4 | 458 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |