Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159571.1 | NP_001153043.1 | 9 - 10 | 2204 | 62 | |
NM_010144.6 | NP_034274.4 | 9 - 10 | 2177 | 62 | ||
GenBank mRNA | AK046884.1 | - | 9 - 10 | 2142 | 62 | |
AK049340.1 | - | 9 - 10 | 2178 | 62 | ||
AK132392.1 | - | 9 - 10 | 2177 | 62 | ||
AK143422.1 | - | 2147 | 62 | |||
BC013342.1 | - | 9 - 10 | 1865 | 62 | ||
BC090839.1 | - | 9 - 10 | 1954 | 62 | ||
U06834.1 | - | 10 - 11 | 1790 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |