Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100030612.mm.1 |
NM_001033711.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030612.mm.1 |
NM_001033711.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030612.mm.1 |
TC1100003326.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030615.mm.1 |
NM_001033711.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030615.mm.1 |
NM_001033711.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030615.mm.1 |
NM_010161.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030615.mm.1 |
NM_010161.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030615.mm.1 |
NM_146023.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030615.mm.1 |
TC1100003326.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030613.mm.1 |
NM_001033711.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030613.mm.1 |
NM_001033711.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030613.mm.1 |
TC1100003326.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030614.mm.1 |
NM_001033711.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030614.mm.1 |
NM_001033711.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030614.mm.1 |
TC1100003326.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030611.mm.1 |
NM_001033711.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030611.mm.1 |
NM_001033711.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030611.mm.1 |
TC1100003326.mm.1 |
MTA TC |