Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010195.2 | NP_034325.2 | 1 - 2 | 496 | 65 | |
GenBank mRNA | AF110818.1 | - | 1 - 2 | 209 | 65 | |
AK047873.1 | - | 1 - 2 | 478 | 65 | ||
AK132387.1 | - | 1 - 2 | 496 | 65 | ||
AK134981.1 | - | 1 - 2 | 477 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mus musculus Probe spans exons |