Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010200.3 | NP_034330.2 | 1 - 2 | 562 | 75 | |
GenBank mRNA | AF020737.1 | - | 1 - 2 | 195 | 75 | |
BC018238.1 | - | 1 - 2 | 562 | 4481123 | 75 | |
U66202.1 | - | 1 - 2 | 189 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |