Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010202.5 | NP_034332.2 | 1 - 2 | 443 | 102 | |
GenBank mRNA | AK082814.1 | - | 1 - 2 | 444 | 102 | |
BC104312.1 | - | 1 - 2 | 324 | 40044462 | 102 | |
BC104313.1 | - | 1 - 2 | 324 | 40044466 | 102 | |
M30642.1 | - | 1 - 2 | 324 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |