Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Figf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC080770.1 | - | 4 - 5 | 911 | 30286444 | 103 |
D89628.1 | - | 4 - 5 | 743 | 103 | ||
X99572.1 | - | 4 - 5 | 930 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |