Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbegf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010415.2 | NP_034545.1 | 1 - 2 | 337 | 62 | |
GenBank mRNA | AK052084.1 | - | 1 - 2 | 308 | 62 | |
AK145548.1 | - | 1 - 2 | 306 | 62 | ||
AK160575.1 | - | 1 - 2 | 307 | 62 | ||
AK164309.1 | - | 1 - 2 | 338 | 62 | ||
BC089607.1 | - | 1 - 2 | 306 | 6751441 | 62 | |
L07264.1 | - | 1 - 2 | 270 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |