Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hipk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136065.2 | NP_001129537.1 | 10 - 11 | 2469 | 73 | |
NM_001294143.1 | NP_001281072.1 | 10 - 11 | 2548 | 73 | ||
NM_001294144.1 | NP_001281073.1 | 10 - 11 | 2545 | 73 | ||
NM_010433.2 | NP_034563.2 | 10 - 11 | 2629 | 73 | ||
GenBank mRNA | AF077659.1 | - | 10 - 11 | 2392 | 73 | |
AF170301.1 | - | 10 - 11 | 2298 | 73 | ||
AF170302.1 | - | 10 - 11 | 2379 | 73 | ||
AF208292.1 | - | 10 - 11 | 2382 | 73 | ||
AF273680.1 | - | 10 - 11 | 2269 | 73 | ||
AF333791.1 | - | 10 - 11 | 2172 | 73 | ||
AF333792.1 | - | 10 - 11 | 2256 | 73 | ||
BC031904.1 | - | 10 - 11 | 2281 | 5368577 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |