Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010575.2 | NP_034705.2 | 25 - 26 | 2759 | 61 | |
GenBank mRNA | AF045019.1 | - | 2 - 3 | 223 | 61 | |
AF170316.1 | - | 25 - 26 | 2620 | 61 | ||
AK048700.1 | - | 20 - 21 | 2267 | 61 | ||
AK140749.1 | - | 20 - 21 | 2411 | 61 | ||
AK142289.1 | - | 25 - 26 | 2621 | 61 | ||
AK154619.1 | - | 25 - 26 | 2760 | 61 | ||
AK156872.1 | - | 24 - 25 | 2940 | 61 | ||
AK157012.1 | - | 24 - 25 | 2944 | 61 | ||
AK158443.1 | - | 25 - 26 | 2611 | 61 | ||
AK161727.1 | - | 4 - 5 | 635 | 61 | ||
BC120493.1 | - | 25 - 26 | 2609 | 40129416 | 61 | |
BC137570.1 | - | 25 - 26 | 2609 | 8860589 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |