Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010575.2 | NP_034705.2 | 28 - 29 | 3098 | 65 | |
GenBank mRNA | AF045019.1 | - | 5 - 6 | 562 | 65 | |
AF170316.1 | - | 28 - 29 | 2959 | 65 | ||
AK140749.1 | - | 23 - 24 | 2750 | 65 | ||
AK142289.1 | - | 28 - 29 | 2960 | 65 | ||
AK154619.1 | - | 28 - 29 | 3099 | 65 | ||
AK156872.1 | - | 27 - 28 | 3279 | 65 | ||
AK157012.1 | - | 27 - 28 | 3283 | 65 | ||
AK158443.1 | - | 28 - 29 | 2950 | 65 | ||
BC120493.1 | - | 28 - 29 | 2948 | 40129416 | 65 | |
BC137570.1 | - | 28 - 29 | 2948 | 8860589 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |