Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010612.2 | NP_034742.2 | 2 - 3 | 423 | 99 | |
GenBank mRNA | AK054510.1 | - | 2 - 3 | 456 | 99 | |
AK141938.1 | - | 2 - 3 | 446 | 99 | ||
BC020530.1 | - | 2 - 3 | 423 | 4238984 | 99 | |
EU884114.1 | - | 2 - 3 | 453 | 99 | ||
S53103.1 | - | 2 - 3 | 434 | 99 | ||
X59397.1 | - | 2 - 3 | 370 | 99 | ||
X70842.1 | - | 2 - 3 | 448 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |