Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010612.2 | NP_034742.2 | 11 - 12 | 1791 | 91 | |
GenBank mRNA | BC020530.1 | - | 11 - 12 | 1791 | 4238984 | 91 |
EU884114.1 | - | 11 - 12 | 1821 | 91 | ||
S53103.1 | - | 11 - 12 | 1802 | 91 | ||
X59397.1 | - | 11 - 12 | 1738 | 91 | ||
X70842.1 | - | 11 - 12 | 1816 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |