Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252658.1 | NP_001239587.1 | 11 - 12 | 1902 | 90 | |
NM_001252659.1 | NP_001239588.1 | 11 - 12 | 1899 | 90 | ||
NM_010700.3 | NP_034830.2 | 11 - 12 | 1902 | 90 | ||
GenBank mRNA | AK149950.1 | - | 11 - 12 | 1840 | 90 | |
AK151968.1 | - | 11 - 12 | 1869 | 90 | ||
AK152103.1 | - | 11 - 12 | 1838 | 90 | ||
AK160006.1 | - | 11 - 12 | 1840 | 90 | ||
BC053041.1 | - | 11 - 12 | 1847 | 5704871 | 90 | |
X64414.1 | - | 11 - 12 | 1821 | 90 | ||
Z19521.1 | - | 11 - 12 | 1700 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |