Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010730.2 | NP_034860.2 | 3 - 4 | 249 | 82 | |
GenBank mRNA | AK002681.1 | - | 3 - 4 | 251 | 82 | |
AK140838.1 | - | 3 - 4 | 252 | 82 | ||
AK150564.1 | - | 3 - 4 | 251 | 82 | ||
AK150718.1 | - | 3 - 4 | 251 | 82 | ||
AK153156.1 | - | 3 - 4 | 251 | 82 | ||
AK153490.1 | - | 3 - 4 | 251 | 82 | ||
AK167612.1 | - | 3 - 4 | 251 | 82 | ||
BC002289.1 | - | 2 - 3 | 204 | 3590168 | 82 | |
BC004594.1 | - | 2 - 3 | 190 | 3587973 | 82 | |
CT009771.1 | - | 2 - 3 | 175 | 82 | ||
CT010298.1 | - | 2 - 3 | 175 | 82 | ||
EU684130.1 | - | 2 - 3 | 175 | 82 | ||
M24554.1 | - | 2 - 3 | 160 | 82 | ||
X07486.1 | - | 2 - 3 | 188 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |