Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039543.2 | NP_001034632.1 | 5 - 6 | 572 | 93 | |
NM_010801.3 | NP_034931.2 | 4 - 5 | 527 | 93 | ||
GenBank mRNA | AF009515.1 | - | 4 - 5 | 468 | 93 | |
AF100171.1 | - | 4 - 5 | 468 | 93 | ||
AK007292.1 | - | 4 - 5 | 526 | 93 | ||
AK133448.1 | - | 4 - 5 | 528 | 93 | ||
BC027526.1 | - | 5 - 6 | 521 | 1495721 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |