Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010927.3 | NP_035057.1 | 8 - 9 | 1028 | 117 | |
GenBank mRNA | AF065919.1 | - | 8 - 9 | 1025 | 117 | |
AF065920.1 | - | 8 - 9 | 1025 | 117 | ||
AF065921.2 | - | 8 - 9 | 1025 | 117 | ||
AF065922.2 | - | 7 - 8 | 898 | 117 | ||
AF065923.2 | - | 7 - 8 | 898 | 117 | ||
AY090567.1 | - | 7 - 8 | 848 | 117 | ||
BC062378.1 | - | 8 - 9 | 1037 | 4978648 | 117 | |
M84373.1 | - | 8 - 9 | 1029 | 117 | ||
M87039.1 | - | 8 - 9 | 1100 | 117 | ||
M92649.1 | - | 8 - 9 | 1036 | 117 | ||
U43428.1 | - | 8 - 9 | 916 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |