Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011102.4 | NP_035232.1 | 8 - 9 | 1164 | 79 | |
GenBank mRNA | AK144419.1 | - | 8 - 9 | 1166 | 79 | |
L28035.1 | - | 8 - 9 | 1242 | 79 | ||
X67129.1 | - | 8 - 9 | 973 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |