Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291434.1 | NP_001278363.1 | 15 - 16 | 1870 | 60 | |
NM_011102.4 | NP_035232.1 | 16 - 17 | 2023 | 60 | ||
GenBank mRNA | AK144419.1 | - | 16 - 17 | 2025 | 60 | |
BC111807.1 | - | 15 - 16 | 1757 | 40039740 | 60 | |
L28035.1 | - | 16 - 17 | 2101 | 60 | ||
X67129.1 | - | 16 - 17 | 1832 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |