Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011182.4 | NP_035312.3 | 1 - 2 | 233 | 90 | |
GenBank mRNA | AB013470.1 | - | 1 - 2 | 87 | 90 | |
AF084221.1 | - | 1 - 2 | 127 | 90 | ||
AK036191.1 | - | 1 - 2 | 147 | 90 | ||
AK142407.1 | - | 1 - 2 | 222 | 90 | ||
AK159229.1 | - | 1 - 2 | 205 | 90 | ||
AK168566.1 | - | 1 - 2 | 147 | 90 | ||
AK220565.1 | - | 1 - 2 | 213 | 90 | ||
BC035296.1 | - | 1 - 2 | 174 | 3969080 | 90 | |
BY271755.1 | - | 1 - 2 | 233 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |