Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pth1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083935.1 | NP_001077404.1 | 9 - 10 | 1266 | 96 | |
NM_001083936.1 | NP_001077405.1 | 8 - 9 | 1299 | 96 | ||
NM_011199.2 | NP_035329.2 | 10 - 11 | 1268 | 96 | ||
GenBank mRNA | BC013446.1 | - | 10 - 11 | 1245 | 4241234 | 96 |
BC051981.1 | - | 8 - 9 | 1170 | 6402767 | 96 | |
X78936.1 | - | 8 - 9 | 1045 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |