Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scnn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272023.1 | NP_001258952.1 | 2 - 3 | 417 | 92 | |
NM_011325.2 | NP_035455.1 | 2 - 3 | 414 | 92 | ||
NR_073548.1 | 2 - 3 | 414 | 92 | |||
GenBank mRNA | AF112186.1 | - | 2 - 3 | 341 | 92 | |
AK028909.1 | - | 2 - 3 | 396 | 92 | ||
AK036819.1 | - | 2 - 3 | 415 | 92 | ||
AK087055.1 | - | 2 - 3 | 376 | 92 | ||
AK164709.1 | - | 2 - 3 | 377 | 92 | ||
BC131969.1 | - | 2 - 3 | 330 | 40130246 | 92 | |
BC132397.1 | - | 2 - 3 | 330 | 40130674 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |