Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011404.3 | NP_035534.2 | 4 - 5 | 911 | 71 | |
GenBank mRNA | AB017189.1 | - | 4 - 5 | 857 | 71 | |
AB023409.1 | - | 4 - 5 | 839 | 71 | ||
AK142288.1 | - | 4 - 5 | 912 | 71 | ||
AK148134.1 | - | 4 - 5 | 581 | 71 | ||
AK192999.1 | - | 2 - 3 | 70 | 71 | ||
AK202552.1 | - | 2 - 3 | 70 | 71 | ||
AK215643.1 | - | 2 - 3 | 100 | 71 | ||
BC026131.1 | - | 4 - 5 | 920 | 4188854 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |