Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011530.3 | NP_035660.3 | 2 - 3 | 641 | 65 | |
GenBank mRNA | AK020022.1 | - | 2 - 3 | 554 | 65 | |
AK145234.1 | - | 2 - 3 | 65 | |||
AK151376.1 | - | 2 - 3 | 551 | 65 | ||
AK151483.1 | - | 2 - 3 | 551 | 65 | ||
AK151657.1 | - | 2 - 3 | 574 | 65 | ||
AK153287.1 | - | 2 - 3 | 551 | 65 | ||
AK159200.1 | - | 2 - 3 | 607 | 65 | ||
AK161091.1 | - | 2 - 3 | 555 | 65 | ||
BC005578.1 | - | 2 - 3 | 541 | 3968225 | 65 | |
BC051257.1 | - | 2 - 3 | 554 | 6530488 | 65 | |
BC057668.1 | - | 2 - 3 | 927 | 65 | ||
KJ650229.1 | - | 1 - 2 | 491 | 65 | ||
M90459.1 | - | 1 - 2 | 488 | 65 | ||
U60087.1 | - | 1 - 2 | 491 | 65 | ||
U60088.1 | - | 1 - 2 | 491 | 65 | ||
U60089.1 | - | 1 - 2 | 491 | 65 | ||
U60090.1 | - | 1 - 2 | 65 | |||
U60091.1 | - | 1 - 2 | 491 | 65 | ||
U60092.1 | - | 1 - 2 | 488 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |