Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011530.3 | NP_035660.3 | 6 - 7 | 1299 | 80 | |
GenBank mRNA | AK079667.1 | - | 6 - 7 | 1213 | 80 | |
AK145234.1 | - | 6 - 7 | 1259 | 80 | ||
AK151376.1 | - | 6 - 7 | 1209 | 80 | ||
AK151483.1 | - | 6 - 7 | 1209 | 80 | ||
AK151657.1 | - | 6 - 7 | 1232 | 80 | ||
AK153287.1 | - | 6 - 7 | 1209 | 80 | ||
AK159200.1 | - | 6 - 7 | 1265 | 80 | ||
AK161091.1 | - | 6 - 7 | 1215 | 80 | ||
AK169645.1 | - | 3 - 4 | 507 | 80 | ||
BC005578.1 | - | 6 - 7 | 1199 | 3968225 | 80 | |
BC051257.1 | - | 6 - 7 | 1212 | 6530488 | 80 | |
BC057668.1 | - | 6 - 7 | 1586 | 80 | ||
CT010387.1 | - | 5 - 6 | 1146 | 80 | ||
KJ650229.1 | - | 5 - 6 | 1149 | 80 | ||
M90459.1 | - | 5 - 6 | 1146 | 80 | ||
U60087.1 | - | 5 - 6 | 1149 | 80 | ||
U60088.1 | - | 5 - 6 | 1149 | 80 | ||
U60089.1 | - | 5 - 6 | 1149 | 80 | ||
U60090.1 | - | 5 - 6 | 1149 | 80 | ||
U60091.1 | - | 5 - 6 | 1149 | 80 | ||
U60092.1 | - | 5 - 6 | 1146 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |