Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tie1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011587.2 | NP_035717.2 | 12 - 13 | 2060 | 110 | |
GenBank mRNA | AK051512.1 | - | 12 - 13 | 2006 | 110 | |
AK052192.1 | - | 12 - 13 | 1980 | 110 | ||
AK052413.1 | - | 12 - 13 | 1993 | 110 | ||
AK161898.1 | - | 12 - 13 | 1979 | 110 | ||
BC046452.1 | - | 12 - 13 | 1982 | 4195879 | 110 | |
X71425.1 | - | 12 - 13 | 1961 | 110 | ||
X73960.1 | - | 12 - 13 | 1960 | 110 | ||
X80764.1 | - | 12 - 13 | 2274 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |