Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011594.3 | NP_035724.2 | 2 - 3 | 558 | 85 | |
GenBank mRNA | AK032818.1 | - | 2 - 3 | 472 | 85 | |
AK081522.1 | - | 2 - 3 | 654 | 85 | ||
AK144236.1 | - | 2 - 3 | 473 | 85 | ||
AK194058.1 | - | 1 - 2 | 112 | 85 | ||
AY622854.1 | - | 1 - 2 | 88 | 85 | ||
BC049126.1 | - | 2 - 3 | 455 | 6821553 | 85 | |
M82858.1 | - | 2 - 3 | 453 | 85 | ||
X62622.1 | - | 2 - 3 | 463 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |