Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011596.5 | NP_035726.2 | 2174 | 57 | ||
GenBank mRNA | AB022323.1 | - | 2140 | 57 | ||
AF218252.1 | - | 2159 | 57 | |||
AF388674.1 | - | 3 - 4 | 348 | 57 | ||
AK017467.1 | - | 2 - 3 | 198 | 57 | ||
AK032909.1 | - | 2 - 3 | 928 | 57 | ||
AK155055.1 | - | 2151 | 57 | |||
AK170122.1 | - | 2671 | 57 | |||
BC018369.1 | - | 2127 | 57 | |||
BC108991.1 | - | 2071 | 40047832 | 57 | ||
BC108992.1 | - | 2071 | 40047833 | 57 | ||
BC112905.1 | - | 2134 | 3670722 | 57 | ||
M31226.1 | - | 2070 | 57 | |||
X55184.1 | - | 2071 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |