Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039363.2 | NP_001034452.1 | 2 - 3 | 255 | 79 | |
NM_001286713.1 | NP_001273642.1 | 2 - 3 | 255 | 79 | ||
NM_001286714.1 | NP_001273643.1 | 2 - 3 | 250 | 79 | ||
NM_001286716.1 | NP_001273645.1 | 2 - 3 | 250 | 79 | ||
NM_001286718.1 | NP_001273647.1 | 2 - 3 | 255 | 79 | ||
NM_001286720.1 | NP_001273649.1 | 2 - 3 | 255 | 79 | ||
NM_011647.3 | NP_035777.2 | 2 - 3 | 255 | 79 | ||
GenBank mRNA | AK143799.1 | - | 2 - 3 | 256 | 79 | |
AK147016.1 | - | 2 - 3 | 231 | 79 | ||
AK147487.1 | - | 2 - 3 | 224 | 79 | ||
AK147874.1 | - | 2 - 3 | 220 | 79 | ||
AK148062.1 | - | 2 - 3 | 247 | 79 | ||
AK170589.1 | - | 2 - 3 | 246 | 79 | ||
AK171007.1 | - | 2 - 3 | 227 | 79 | ||
BC052449.1 | - | 2 - 3 | 231 | 6409037 | 79 | |
BC060701.1 | - | 2 - 3 | 231 | 6409037 | 79 | |
BY742714.1 | - | 2 - 3 | 259 | 79 | ||
U37775.1 | - | 1 - 2 | 170 | 79 | ||
U39818.1 | - | 2 - 3 | 226 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |