Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011832.2 | NP_035962.2 | 10 - 11 | 2647 | 67 | |
GenBank mRNA | AB007135.1 | - | 10 - 11 | 2536 | 67 | |
BC137855.1 | - | 10 - 11 | 2504 | 8860874 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |