Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159500.1 | NP_001152972.1 | 2 - 3 | 835 | 57 | |
NM_011934.4 | NP_036064.3 | 2 - 3 | 734 | 57 | ||
GenBank mRNA | AK032025.1 | - | 2 - 3 | 542 | 57 | |
AK044339.1 | - | 2 - 3 | 735 | 57 | ||
AK052256.1 | - | 2 - 3 | 837 | 57 | ||
BC044858.1 | - | 1 - 2 | 404 | 57 | ||
BC132595.1 | - | 2 - 3 | 699 | 40130872 | 57 | |
BC132597.1 | - | 2 - 3 | 700 | 40130874 | 57 | |
S82458.1 | - | 1 - 2 | 442 | 57 | ||
X89594.1 | - | 1 - 2 | 442 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons |