Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256042.1 | NP_001242971.1 | 1 - 2 | 196 | 62 | |
NM_011939.3 | NP_036069.1 | 1 - 2 | 196 | 62 | ||
NR_045689.1 | 1 - 2 | 196 | 62 | |||
GenBank mRNA | AB029349.1 | - | 1 - 2 | 128 | 62 | |
AB029350.1 | - | 1 - 2 | 123 | 62 | ||
AF160966.1 | - | 1 - 2 | 123 | 62 | ||
BC138130.1 | - | 1 - 2 | 196 | 8861149 | 62 | |
BC145153.1 | - | 1 - 2 | 196 | 62 | ||
BC145154.1 | - | 1 - 2 | 196 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |