Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF139923.1 | - | 5 - 6 | 1439 | 66 | |
AK135153.1 | - | 5 - 6 | 1628 | 66 | ||
AK137418.1 | - | 6 - 7 | 1764 | 66 | ||
BC117995.1 | - | 5 - 6 | 1439 | 40096022 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |