Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013482.2 | NP_038510.2 | 8 - 9 | 938 | 75 | |
GenBank mRNA | BC053392.1 | - | 8 - 9 | 874 | 30075322 | 75 |
L08967.1 | - | 8 - 9 | 914 | 75 | ||
L10627.1 | - | 8 - 9 | 949 | 75 | ||
L29788.1 | - | 8 - 9 | 910 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |