Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013485.1 | NP_038513.1 | 8 - 9 | 1308 | 100 | |
GenBank mRNA | BC011137.1 | - | 8 - 9 | 1308 | 4195787 | 100 |
X05475.1 | - | 8 - 9 | 1287 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |