Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039195.1 | NP_001034284.1 | 9 - 10 | 1675 | 94 | |
NM_001083806.1 | NP_001077275.1 | 9 - 10 | 1675 | 94 | ||
NM_013540.2 | NP_038568.2 | 9 - 10 | 1675 | 94 | ||
GenBank mRNA | AB109210.1 | - | 9 - 10 | 1268 | 94 | |
AB109211.1 | - | 9 - 10 | 1268 | 94 | ||
AB111957.1 | - | 9 - 10 | 1268 | 94 | ||
AK014389.1 | - | 9 - 10 | 1720 | 94 | ||
AK043490.1 | - | 9 - 10 | 1648 | 94 | ||
AK044574.1 | - | 9 - 10 | 1607 | 94 | ||
AK046861.1 | - | 9 - 10 | 1675 | 94 | ||
AK086614.1 | - | 9 - 10 | 1609 | 94 | ||
AK134572.1 | - | 9 - 10 | 1605 | 94 | ||
AK134647.1 | - | 8 - 9 | 1121 | 94 | ||
AK134772.1 | - | 8 - 9 | 1246 | 94 | ||
AK141525.1 | - | 9 - 10 | 1651 | 94 | ||
AK163941.1 | - | 9 - 10 | 1668 | 94 | ||
BC048248.1 | - | 9 - 10 | 1534 | 6494013 | 94 | |
BC066193.1 | - | 9 - 10 | 1463 | 94 | ||
X57498.1 | - | 9 - 10 | 1470 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |