Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013559.2 | NP_038587.2 | 9 - 10 | 1487 | 61 | |
GenBank mRNA | AK083179.1 | - | 1348 | 61 | ||
AK146697.1 | - | 9 - 10 | 1406 | 61 | ||
AK160824.1 | - | 9 - 10 | 1468 | 61 | ||
AK165046.1 | - | 9 - 10 | 1455 | 61 | ||
AK172913.1 | - | 3 - 4 | 384 | 61 | ||
BC018378.1 | - | 9 - 10 | 1362 | 3257042 | 61 | |
D67016.1 | - | 9 - 10 | 1302 | 61 | ||
D67017.1 | - | 9 - 10 | 1389 | 61 | ||
L40406.1 | - | 9 - 10 | 1387 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |