Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lifr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113386.1 | NP_001106857.1 | 8 - 9 | 1387 | 109 | |
NM_013584.2 | NP_038612.1 | 7 - 8 | 1036 | 109 | ||
GenBank mRNA | AK145864.1 | - | 8 - 9 | 1388 | 109 | |
AK155524.1 | - | 7 - 8 | 1263 | 109 | ||
BC031929.1 | - | 7 - 8 | 1029 | 4159053 | 109 | |
D17444.1 | - | 7 - 8 | 1043 | 109 | ||
D26177.1 | - | 7 - 8 | 1043 | 109 | ||
S73495.1 | - | 7 - 8 | 1043 | 109 | ||
S73496.1 | - | 7 - 8 | 1043 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Mouse Stem Cell Pluripotency Inventoried Mouse_Stem_Cell Probe spans exons |