Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013657.5 | NP_038685.3 | 9 - 10 | 1102 | 71 | |
GenBank mRNA | AK034239.1 | - | 9 - 10 | 1161 | 71 | |
AK140348.1 | - | 1638 | 71 | |||
BC066852.1 | - | 9 - 10 | 1102 | 30093230 | 71 | |
X85994.1 | - | 9 - 10 | 1054 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |