Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163489.1 | NP_001156961.1 | 12 - 13 | 1497 | 69 | |
NM_001163490.1 | NP_001156962.1 | 12 - 13 | 1576 | 69 | ||
NM_001163491.1 | NP_001156963.1 | 12 - 13 | 1535 | 69 | ||
NM_013658.3 | NP_038686.3 | 12 - 13 | 1675 | 69 | ||
GenBank mRNA | AK035918.1 | - | 11 - 12 | 1537 | 69 | |
AK077976.1 | - | 12 - 13 | 1656 | 69 | ||
AK152728.1 | - | 12 - 13 | 1667 | 69 | ||
AK153164.1 | - | 12 - 13 | 1667 | 69 | ||
AK154392.1 | - | 12 - 13 | 1636 | 69 | ||
BC025800.1 | - | 12 - 13 | 1593 | 5102015 | 69 | |
X85991.1 | - | 12 - 13 | 1497 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |